Order Entry
Belgium
ContactUsLinkComponent
 
Potassium dichromate solution for UV spectrophotometry
Catalog # ACSDIP220
Supplier:  CPACHEM
New Product
CAS Number:  
Potassium dichromate solution for UV spectrophotometry
Catalog # ACSDIP220
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    80 mL
  • Seq:
    2425

Specifications

About this item

 :
 :294.18 g/mol
Boiling Pt:100 °C
Density:~1,00143 g/cm³
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367