Order Entry
Belgium
ContactUsLinkComponent
Mouse Glycogen Synthase Kinase ELISA Kit
Mouse Glycogen Synthase Kinase ELISA Kit
  ANTIA78209-96
New Product
 :  
undefined
Mouse Glycogen Synthase Kinase ELISA Kit
  ANTIA78209-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Glycogen Synthase Kinase
  • Description:
    Mouse Glycogen Synthase Kinase ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Glycogen Synthase Kinase ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Glycogen Synthase Kinase. No significant cross-reactivity or interference between Glycogen Synthase Kinase and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,313 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,188 ng/ml
  • Regulatory status:
    RUO

 

 

Mouse Glycogen Synthase Kinase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Glycogen Synthase Kinase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Glycogen Synthase Kinase ELISA kit (A78209) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Glycogen Synthase Kinase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Glycogen Synthase Kinase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Glycogen Synthase Kinase present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Glycogen Synthase Kinase Antibody, which binds the captured Glycogen Synthase Kinase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Glycogen Synthase Kinase captured in each well. The concentration of Glycogen Synthase Kinase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.