Order Entry
Belgium
ContactUsLinkComponent
L(+)-Arginine 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L(+)-Arginine 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
Certificates
undefined
L(+)-Arginine 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Specification Test Results

  • USP - Assay (C6Hl4N402) (dried basis):
    98.5 - 101.5%
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation (A]25 AO (+):
    26.3 - 27.7°
  • USP - Loss on Orying at 105 °C:
    ≤0.5%
  • USP - Chloride (Cl):
    ≤0.05%
  • USP - Residue on Ignition:
    ≤0.3%
  • USP - Sulfate (S04):
    ≤0.03%
  • USP - Iron (Fe):
    ≤0.003%
  • USP - Related Compounds - Individual Impurities:
    ≤0.5%
  • USP - Related Compounds - Total Impurities:
    ≤2.0%
  • EP/BP - Assay (C6Hl4N402) (dried basis):
    98.5 - 101.0%
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation (A]2°AO (+):
    25.5 - 28.5 °
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤0.2%
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤0.5%
  • EP/BP - Chloride (Cl):
    ≤200ppm
  • EP/BP - Sulfate (S04):
    ≤300 ppm
  • EP/BP - Ammonium (NH4):
    ≤0.02%
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Orying:
    ≤0.5%
  • EP/BP - Ash (sulfated):
    ≤0.1%
  • JP - Identification:
    Passes Test
  • JP - 0ptical rotation, 20 °C (+):
    26.9 - 27.9°
  • JP - pH (1:10):
    10.5 - 12.0
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.021%
  • JP - Sulfate (S04):
    ≤0.028%
  • JP - Ammonium (NH4):
    ≤0.02%
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Iron (Fe):
    ≤10 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Orying at 105 °C:
    ≤0.30%
  • JP - Residue after Ignition:
    ≤0.1%
  • JP - Assay (C6Hl4N402)(dried basis):
    98.5 - 101.0%
  • ChP - Specific Rotation @ 20 °C:
    26.9 - 27.9
  • ChP - Identification 1 (TLC):
    Passes Test
  • ChP - Identification 2 (IR):
    Passes Test
  • ChP - Alkalinity (pH):
    10.5 - 12.0
  • ChP - Transmittance of solution:
    0.98
  • ChP - Chlorides:
    ≤0.02%
  • ChP - Sulfates:
    ≤0.02%
  • ChP - Ammonium:
    ≤0.02%
  • ChP - Proteins:
    Passes Test
  • ChP - 0ther amino acids (TLC)(<=0.4%, one spot only):
    Passes Test
  • ChP - Loss on drying @ 105 °C:
    ≤0.5%
  • ChP - Residue on ignition:
    ≤0.1%
  • ChP - Iron:
    ≤0.001%
  • ChP - Heavy metals:
    ≤0.001%
  • ChP - Arsenic:
    ≤0.0001%
  • ChP - Assay, dried basis:
    99.0 - 101.0%

Specification Test Results

Related Information

Frequently Bought Together