Order Entry
Belgium
ContactUsLinkComponent
Ammonium carbonate USP Test Solution (TS)
Irritant
Ammonium carbonate USP Test Solution (TS)
Catalog # REGEUSP0605
Supplier:  REAGECON
New Product
CAS Number:  
Irritant
Ammonium carbonate USP Test Solution (TS)
Catalog # REGEUSP0605
Supplier:  REAGECON
New Product
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle GL45
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :96.09 g/mol
 :GHS07
CAS Number:506-87-6
 :10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890