Order Entry
Belgium
ContactUsLinkComponent
Soyabean Casein Digest Medium
Soyabean Casein Digest Medium
Catalog # HIMELQ193CC-5X200M
Supplier:  HIMEDIA
New Product
CAS Number:  
Soyabean Casein Digest Medium
Catalog # HIMELQ193CC-5X200M
Supplier:  HIMEDIA
New Product
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Description:
    Soyabean casein digest medium with 0,5% soya lecithin and 4% Tween 20
  • Storage conditions:
    Store between 15...30 °C. Use before expiry date on the label. Product performance is best if used within stated expiry period.
  • pH:
    6,80- 7,80

Specifications

About this item

Sterility test medium also used for cultivation of soyabean casein digest medium.

  • Colour: Light yellow coloured opalascent solution
  • Appearance: Sterile soyabean casein digest medium with 0,5% Soya Lecithin and 4% Tween 20 in glass bottle
  • Quantity of medium: 200 ml of medium in glass bottle
  • Cultural response: Cultural characteristics observed after an incubation as specified

Soyabean casein digest medium with 0,5% Soya Lecithin and 4% Tween 20 is used for the detection of microorganisms on surfaces of sanitary importance. Tryptone and soya peptone makes this medium nutritious by providing nitrogeneous and carbonaceous compounds, long chain peptides, vitamins and other essential nutrients for the growth of microorganisms.

Caution: Read the label before opening the container. Wear protective gloves/protective clothing/eye protection/face protection. Follow good microbiological lab practices while handling specimens and culture. Standard precautions as per established guidelines should be followed while handling specimens.