Order Entry
Belgium
Orders LinkContactUsLinkComponent
TOC Standard Solution

TOC Standard Solution

TOC standard solution, 1000 mg/L, 500 mL
Catalog # CMPLTOC1000.L5
TOC Standard Solution
Catalog # CMPLTOC1000.L5
CAS Number:  
TOC Standard Solution

TOC standard solution, 1000 mg/L, 500 mL

CAS Number:  
Price
Demander un prix
Connexion Pour la disponibilité
1 * 500 mL
Product Details & Documents
Spécifications

Spécifications

  • Pk:
    500 mL
  • Description:
    TOC standard solution, 1000 mg/L
  • Cat. No.:
    CMPLTOC1000.L5
  • Shipping conditions:
    48 months - Store under normal laboratory conditions, at temperatures between 15...25 °C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.