Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dimetridazole

Dimetridazole

250 mg
Catalog # ACSDSB32100.250MG
Irritant
Supplier:   CPACHEM
Dimetridazole
Catalog # ACSDSB32100.250MG
Supplier:  CPACHEM
CAS Number:  551-92-8
Dimetridazole

250 mg

CAS Number:  551-92-8
Price
69,80 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  1,2-Dimethyl-5-nitroimidazole
  • CAS Number:551-92-8
  • Formula:C₅H₇N₃O₂
  • MDL Number:MFCD00047046
  • Molecular Weight:141.13 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:209-001-2
Specifications

Specifications

  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.