Order Entry
Belgium
ContactUsLinkComponent
 
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (Alexa Fluor® 488)
 
undefined
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (Alexa Fluor® 488)

 

  • Catalog No:
  • STMC60109AD.1
  • STMC60109AD
  • Antigen symbol:
  • CD62L
  • CD62L
  • Antigen name:
  • L-Selectin
  • L-Selectin
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • MEL-14
  • MEL-14
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 20343
  • 20343
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

 

Frequently Bought Together