Order Entry
Belgium
ContactUsLinkComponent
Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530] (PE)
Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530] (PE)
Certificates
undefined
Anti-Igg2b Isotype Control Rat Monoclonal Antibody [clone: RTK4530] (PE)

Specifications

  • Catalog No:
  • STMC60077PE.1
  • STMC60077PE
  • STMC60077PE.3
  • STMC60077PE.2
  • Antigen name:
  • Igg2b Isotype Control
  • Igg2b Isotype Control
  • Igg2b Isotype Control
  • Igg2b Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • RTK4530
  • RTK4530
  • RTK4530
  • RTK4530
  • Host:
  • Rat
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together