Mouse monoclonal IgG3 antibody against human, mouse, rat SSEA-4, FITC-conjugated.
- Pluripotent Stem cells
The MC-813-70 antibody reacts with stage-specific embryonic antigen-4 (SSEA-4), a glycolipid carbohydrate antigen expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES) and induced pluripotent stem (iPS) cells, a subset of mesenchymal stem cells, and rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG, ES, and iPS cells. Expression of SSEA-4 is down-regulated following differentiation of human EC, ES, and iPS cells. In contrast, the differentiation of mouse EC, ES, or iPS cells may be accompanied by an increase in SSEA-4 expression. This antibody clone has been verified for labeling human ES and iPS cells grown in mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860), and has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog #18166) and EasySep™ hESC/hiPSC SSEA-4 Positive Selection Kit (Catalog #18165; partial blocking may be observed).
Type:
Antigen: SSEA-4
Clonality: Monoclonal
Clone: MC-813-70
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Cat;Chicken;Dog;Human;Mouse;Rabbit;Rat;Rhesus monkey
- Catalog No:
- STMC60062FI.1
- STMC60062FI
- Antigen name:
- SSEA-4
- SSEA-4
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- MC-813-70
- MC-813-70
- Reactivity:
- Rabbit, Rhesus monkey, Chicken, Human, Dog, Rat, Mouse, Cat
- Rabbit, Rhesus monkey, Chicken, Human, Dog, Rat, Mouse, Cat
- Host:
- Mouse
- Mouse
- Gene ID:
- 330401
- 330401
- Isotype:
- IgG3 kappa
- IgG3 kappa