Order Entry
Belgium
ContactUsLinkComponent
 
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (FITC)
 
undefined
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (FITC)

 

  • Catalog No:
  • STMC60110FI
  • STMC60110FI.1
  • Antigen name:
  • CD123 (IL-3Ra)
  • CD123 (IL-3Ra)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 6H6
  • 6H6
  • Reactivity:
  • Sooty mangabey, Rhesus monkey, Human
  • Sooty mangabey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3563
  • 3563
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together