Order Entry
Belgium
ContactUsLinkComponent
Anti-NK1.1 (CD161) Mouse Monoclonal Antibody [clone: PK136] (Alexa Fluor® 488)
Anti-NK1.1 (CD161) Mouse Monoclonal Antibody [clone: PK136] (Alexa Fluor® 488)
Certificates
undefined
Anti-NK1.1 (CD161) Mouse Monoclonal Antibody [clone: PK136] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60103AD.1
  • STMC60103AD
  • Antigen name:
  • NK1.1 (CD161)
  • NK1.1 (CD161)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • PK136
  • PK136
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 17059
  • 17059
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together