Order Entry
Belgium
ContactUsLinkComponent
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 3A2A20] (Alexa Fluor® 488)
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 3A2A20] (Alexa Fluor® 488)
Certificates
undefined
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 3A2A20] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60093AD.1
  • STMC60093AD
  • Antigen name:
  • OCT4 (OCT3)
  • OCT4 (OCT3)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • 3A2A20
  • 3A2A20
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5460
  • 5460
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

Specifications

Frequently Bought Together