Order Entry
Belgium
ContactUsLinkComponent
Anti-Sca1 (Ly-6A/E) Rat Monoclonal Antibody [clone: E13-161.7] (PE)
Anti-Sca1 (Ly-6A/E) Rat Monoclonal Antibody [clone: E13-161.7] (PE)
Certificates
undefined
Anti-Sca1 (Ly-6A/E) Rat Monoclonal Antibody [clone: E13-161.7] (PE)

Specifications

  • Catalog No:
  • STMC60032PE.1
  • STMC60032PE
  • Antigen name:
  • Sca1 (Ly-6A/E)
  • Sca1 (Ly-6A/E)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • E13-161.7
  • E13-161.7
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 110454
  • 110454
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together