Order Entry
Belgium
ContactUsLinkComponent
 
Anti-c-KIT Rat Monoclonal Antibody [clone: 2B8] (Alexa Fluor® 488)
 
undefined
Anti-c-KIT Rat Monoclonal Antibody [clone: 2B8] (Alexa Fluor® 488)

 

  • Catalog No:
  • STMC60025AD.1
  • STMC60025AD
  • Antigen symbol:
  • CD117
  • CD117
  • Antigen name:
  • c-KIT
  • c-KIT
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • 2B8
  • 2B8
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16590
  • 16590
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

 

Frequently Bought Together