About this item
Rat monoclonal IgG2b, antibody against mouse MHC Class II (I-A/I-E), FITC-conjugated.
- B cells
- Macrophages
- Monocytes
- T cells
The M5/114.15.2 (M5/114) antibody reacts with an extracellular epitope on the mouse major histocompatibility complex class II receptor (MHC-II). MHC-II is a heterodimeric transmembrane glycoprotein comprising an α subunit associated non-covalently with a β subunit. In mice, MHC-II complexes are designated I-A and I-E and comprise Aα/Aβ and Eα/Eβ heterodimers, respectively. Together with the CD3/T cell receptor (TCR) complex and CD4 molecules, MHC-II mediates a critical function in presenting peptides generated from hydrolysis of exogenous antigens by antigen-presenting cells to CD4+ T (helper) cells, thereby either suppressing or inducing an immune response to the peptides. The M5/114.15.2 antibody recognizes a polymorphic determinant shared by the I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek (but not I-Af, I-Ak, or I-As) MHC class II alloantigens expressed on B cells, dendritic cells, macrophages, monocytes, and activated T cells of mice carrying the H-2b, d, p, q, r, and u haplotypes (but not haplotypes H-2f or H-2s). The M5/114 antibody reportedly inhibits I-A-restricted responses by T cells of the H-2b, d, q, and u (but not H-2f, k, or s) haplotypes. It does not react with cells from NOD (H-2g7) mice.
Type: Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2] (FITC)
Antigen: MHC Class II (I-A/I-E)
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: FITC
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Specifications
- Catalog No:
- STMC100-0294
- STMC100-0296
- STMC100-0295
- Antigen name:
- MHC Class II (I-A/I-E)
- MHC Class II (I-A/I-E)
- MHC Class II (I-A/I-E)
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- M5/114.15.2
- M5/114.15.2
- M5/114.15.2
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 14961/14969
- 14961/14969
- 14961/14969