Mouse monoclonal IgG2a antibody against human, mouse, rat beta-tubulin III, unconjugated.
- Neural cells Psc Derived
- Neurons
The 2G10-TB3 antibody reacts with beta-tubulin III, an ~50 to 55 kDa structural protein that is a constituent of tubulin. Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha- and beta-tubulin subunits. The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types. Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation. It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells. The 2G10-TB3 antibody is expected to recognize all mammalian homologs of beta-tubulin III and the epitope has reportedly been mapped to the extreme C-terminal amino acids, EAQGPK.
Type: Anti-Beta-Tubulin III Mouse Monoclonal Antibody [clone: 2G10-TB3]
Antigen: Beta-Tubulin III
Clonality: Monoclonal
Clone: 2G10-TB3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
- Catalog No:
- STMC60092.1
- STMC60092
- Antigen name:
- Beta-tubulin III
- Beta-Tubulin III
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 2G10-TB3
- 2G10-TB3
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 10381
- 10381
- Isotype:
- IgG2a kappa
- IgG2a kappa