Order Entry
Belgium
ContactUsLinkComponent
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (Alexa Fluor® 488)
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (Alexa Fluor® 488)
Certificates
undefined
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60070AD.1
  • STMC60070AD
  • Antigen name:
  • Igg1 Isotype Control
  • Igg1 Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • MOPC-21
  • MOPC-21
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together