Order Entry
Belgium
ContactUsLinkComponent
Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (APC)
Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (APC)
Certificates
undefined
Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (APC)

Specifications

  • Catalog No:
  • STMC60022AZ
  • STMC60022AZ.1
  • Antigen name:
  • CD8a
  • CD8a
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • RPA-T8
  • RPA-T8
  • Reactivity:
  • Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together