About this item
Mouse monoclonal IgG2b antibody against human CD235ab (glycophorin A/B), FITC-conjugated.
- Red blood cells
The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups. CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types. CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component. The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation. The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes.
Type:
Antigen: CD235ab
Clonality: Monoclonal
Clone: HIR2 (GA-R2)
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- STMC60111FI
- STMC60111FI.1
- Antigen symbol:
- CD235ab
- CD235ab
- Antigen name:
- Glycophorin A/B
- Glycophorin A/B
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- HIR2 (GA-R2)
- HIR2 (GA-R2)
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 2993/2994
- 2993/2994
- Isotype:
- IgG2b kappa
- IgG2b kappa