Order Entry
Belgium
ContactUsLinkComponent
Anti-CD90/CD90.1 Mouse Monoclonal Antibody [clone: OX-7] (FITC)
Anti-CD90/CD90.1 Mouse Monoclonal Antibody [clone: OX-7] (FITC)
Fournisseur:  STEMCELL Technologies
Certificats
undefined
Anti-CD90/CD90.1 Mouse Monoclonal Antibody [clone: OX-7] (FITC)
Fournisseur:  STEMCELL Technologies

Spécifications

  • Catalog No:
  • STMC60024FI
  • STMC60024FI.1
  • Antigen name:
  • CD90/CD90.1
  • CD90/CD90.1
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • OX-7
  • OX-7
  • Reactivity:
  • Rabbit, Guinea pig, Rat, Mouse
  • Rabbit, Guinea pig, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 21838/24832
  • 21838/24832
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Spécifications

Frequently Bought Together