Order Entry
Belgium
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (Alexa Fluor® 488)
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (Alexa Fluor® 488)
Certificates
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (Alexa Fluor® 488)

Specifications

  • Catalog No:
  • STMC60018AD.1
  • STMC60018AD
  • Antigen name:
  • CD45
  • CD45
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • HI30
  • HI30
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together