Specifications
- Antigen name:Perforin
- Clonality:Monoclonal
- Clone:deltaG9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:5551
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody against human perforin, unconjugated.
- Nk cells
- T cells CD8
This mouse monoclonal antibody (clone deltaG9) reacts with human perforin. Perforin is an ~70 kDa protein consisting of four domains: the N- and C-termini, which are related to its function, and two central domains that share similarities to complement proteins C6 - C9. Perforin is involved in the perforin/granzyme apoptosis pathway and is produced by natural killer and cytotoxic T cells. Upon contact with a target cell, perforin monomers are released and aggregate to form pores in the membrane of the target cell, disrupting its membrane integrity. This allows granzyme B to enter the target cell and initiate apoptosis of virally infected and transformed cells.
Type:
Antigen: Perforin
Clonality: Monoclonal
Clone: deltaG9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human