Anti-IgG (H+L) Goat Polyclonal Antibody (iFluor™ 647)
Catalog # STMC100-1081
Supplier: STEMCELL Technologies
Specifications
- Antigen name:IgG (H+L)
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Mouse
- Pk:200 µG
Specifications
About this item
Goat polyclonal antibody against mouse IgG (H+L).
This goat polyclonal antibody reacts with the heavy (H) chains on mouse IgG and the light (L) chains common in most mouse immunoglobulins. Use the polyclonal goat anti-mouse IgG (H+L) antibody with applications, such as immunohistochemistry (IHC) and immunofluorescence (IF) assays. It has been verified for use with immunocytochemistry (ICC).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Type:
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: iFluor™ 647
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse