Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant RANKL
Human Recombinant RANKL
Certificates
undefined
Human Recombinant RANKL

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    CD254|hRANKL2|ODF|OPGL|OPTB2|Osteoclast differentiation factor|soluble Receptor activator of NF-κB ligand|sOdf|TNF-related activation-induced cytokine|TNFSF11|TNF superfamily member 11|TNLG6B|Tumor necrosis factor superfamily member 11|TRANCE
  • Protein/peptide name:
    RANKL
  • Purity:
    ≥95%

Specifications

Frequently Bought Together