Human/Mouse Recombinant BMP-2, ACF
About this item
Bone morphogenetic protein 2 (BMP-2) is a member of the transforming growth factor beta (TGF-β) superfamily. BMP-2 is a disulfide-linked homodimer, acts as a ligand for complexes of type I and II BMP receptors, and primarily activates SMAD1/5/8 signaling (Nohe et al.). BMP-2 is a potent differentiation factor and directs human pluripotent stem cells (hPSCs) towards various cell types including extra-embryonic endoderm, mesenchymal cells, and chondrocytes (Pera et al.). Although BMP-2 expression is low in healthy cartilage, its expression is upregulated at the site of cartilage damage (Blaney Davidson et al.). BMP-2 induces bone and cartilage formation in vitro and is able to induce chondrogenesis in human mesenchymal stem cells (Schmitt et al.). This product is animal component-free.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
3 Options Available Below
Product Details & Documents
Bone morphogenetic protein 2 (BMP-2) is a member of the transforming growth factor beta (TGF-β) superfamily. BMP-2 is a disulfide-linked homodimer, acts as a ligand for complexes of type I and II BMP receptors, and primarily activates SMAD1/5/8 signaling (Nohe et al.). BMP-2 is a potent differentiation factor and directs human pluripotent stem cells (hPSCs) towards various cell types including extra-embryonic endoderm, mesenchymal cells, and chondrocytes (Pera et al.). Although BMP-2 expression is low in healthy cartilage, its expression is upregulated at the site of cartilage damage (Blaney Davidson et al.). BMP-2 induces bone and cartilage formation in vitro and is able to induce chondrogenesis in human mesenchymal stem cells (Schmitt et al.). This product is animal component-free.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- STMC78135
- STMC78135.1
- STMC78135.2
- Protein/peptide name:
- BMP-2, ACF
- BMP-2, ACF
- BMP-2, ACF
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human|Mouse
- Human|Mouse
- Human|Mouse
- Purity:
- ≥95%
- ≥95%
- ≥95%
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.STMC78135Protein/peptide nameBMP-2, ACFProtein/peptide typeRecombinantSpeciesHuman|MousePurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78135.1Protein/peptide nameBMP-2, ACFProtein/peptide typeRecombinantSpeciesHuman|MousePurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78135.2Protein/peptide nameBMP-2, ACFProtein/peptide typeRecombinantSpeciesHuman|MousePurity≥95%Marketsource
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



