Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant SCF (E. coli-expressed)
Human Recombinant SCF (E. coli-expressed)
Certificates
undefined
Human Recombinant SCF (E. coli-expressed)

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    Kit ligand|Mast cell growth factor|Steel factor|Stem cell factor
  • Protein/peptide name:
    SCF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together