Order Entry
Belgium
ContactUsLinkComponent
 
Rat Recombinant GM-CSF
 
undefined
Rat Recombinant GM-CSF

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥95%

 

Frequently Bought Together