Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant PDGF-DD
Human Recombinant PDGF-DD
Certificates
undefined
Human Recombinant PDGF-DD

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    PDGF-DD
  • Purity:
    ≥95%

Specifications

Frequently Bought Together