Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant TNF-beta
Human Recombinant TNF-beta
Fournisseur:  STEMCELL Technologies
Certificats
undefined
Human Recombinant TNF-beta
Fournisseur:  STEMCELL Technologies

Spécifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    TNF-beta
  • Purity:
    ≥95%

Spécifications

Frequently Bought Together