Order Entry
Belgium
ContactUsLinkComponent
Recombinant B18R Protein
Recombinant B18R Protein
Certificates
undefined
Recombinant B18R Protein

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Protein
  • Purity:
    ≥98%

Specifications

Frequently Bought Together