Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant IL-22
Mouse Recombinant IL-22
Certificates
undefined
Mouse Recombinant IL-22

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-22
  • Purity:
    ≥95%

Specifications

Frequently Bought Together