Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant GDF-5
Human Recombinant GDF-5
Catalog # STMC78111.1
CAS Number:  
undefined
Human Recombinant GDF-5
Catalog # STMC78111.1
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    1286
  • Protein/peptide name:
    GDF-5
  • Purity:
    ≥95%

Specifications

About this item

Growth differentiation factor 5 (GDF-5) is a member of the bone morphogenetic protein (BMP) subclass of the transforming growth factor beta (TGF-β) superfamily. It binds a receptor complex comprising BMPR1B and BMPR2, which then activates the SMAD signaling pathway (Carreira et al.; Nishitoh et al.; Osório et al.). GDF-5 enhances chondrogenic differentiation of mesenchymal stem cells, skeletogenesis, and dendrite growth during development (Coleman et al.; Francis-West et al.). Studies in rat demonstrated that GDF-5 regulates patterning, neurogenesis, astrogliogenesis, and neuronal specification (Gajavelli et al.; Krieglstein et al.; O’Keeffe et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').