Order Entry
Belgium
ContactUsLinkComponent
HIV-1 (B Gag) Peptide Pool
HIV-1 (B Gag) Peptide Pool
Catalog # STMC100-1385
CAS Number:  
undefined
HIV-1 (B Gag) Peptide Pool
Catalog # STMC100-1385
CAS Number:  

Specifications

  • Pk:
    1 AU
  • Seq:
    0001
  • Protein/peptide name:
    HIV-1 (B Gag) Peptide Pool
  • Purity:
    Average 70%

Specifications

About this item

Stimulating T cells with HIV-1 (B Gag) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HIV-1 (B Gag) Peptide Pool is a lyophilised mixture of 123 peptides from the gag polyprotein of human immunodeficiency virus 1 (HIV-1). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 500 on gag polyprotein. The HIV-1 gag polyprotein is a key mediator of viral particle assembly (Campbell and Rein; Dong et al.), and together with the endosomal sorting complexes required for transport (ESCRT), results in budding during virion release (Carlson et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').