Order Entry
Belgium
Orders LinkContactUsLinkComponent
STEMdiff™ Cardiomyocyte Freezing Medium

STEMdiff™ Cardiomyocyte Freezing Medium

STEMdiff™ Cardiomyocyte Freezing Medium, 50 ml, Human; animal component-free, 50 mL
Catalog # STMC05030
Marketsource
STEMdiff™ Cardiomyocyte Freezing Medium
Catalog # STMC05030
Marketsource
CAS Number:  
STEMdiff™ Cardiomyocyte Freezing Medium
STEMdiff™ Cardiomyocyte Freezing Medium

STEMdiff™ Cardiomyocyte Freezing Medium, 50 ml, Human; animal component-free, 50 mL

CAS Number:  
Price
195,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

Medium for cryopreserving hPSC-derived cardiomyocytes.

  • Achieve greater than 70% cardiomyocyte viability while maintaining functional capacities
  • Supports hPSC-derived cardiomyocytes generated using the STEMdiff™ Cardiomyocyte differentiation kit

STEMdiff™ Cardiomyocyte freezing medium can be used to cryopreserve cardiomyocytes derived from human pluripotent stem cells (hPSCs) and maintained in STEMdiff™ Cardiomyocyte maintenance medium. STEMdiff™ Cardiomyocyte Dissociation Kit is required for dissociating cardiomyocytes prior to freezing. After thawing using STEMdiff™ Cardiomyocyte support medium, the cardiomyocytes can be used in various downstream applications and analyses. This medium supports hPSC-derived cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte differentiation kit or STEMdiff™ Atrial Cardiomyocyte differentiation kit.

Advantages:
Achieve greater than 70% cardiomyocyte viability while maintaining functional capacities
Supports hPSC-derived cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte differentiation kit or STEMdiff™ Atrial Cardiomyocyte differentiation kit

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    STEMdiff™ Cardiomyocyte Freezing Medium, 50 ml
  • Format:
    Human; animal component-free

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.