Orders LinkContactUsLinkComponent
Ammonium Chloride Solution
Ammonium Chloride Solution
Marketsource
Ammonium Chloride Solution
Marketsource
Ammonium Chloride Solution
Ammonium Chloride Solution
Ammonium chloride solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen or bone marrow cells. It is buffered and optimised for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.
Species: Human, mouse, non human primate, other, rat
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
  • Description
    Pk
  • Reagent for lysis of red blood cells
    100 mL
    1 * 100 mL
    1 * 100 mL - 49,00 €
    49,00 €
    1 * 100 mL
       49,00 €
    Marketsource
    Marketsource
  • Reagent for lysis of red blood cells
    500 mL
    1 * 500 mL
    1 * 500 mL - 152,00 €
    152,00 €
    1 * 500 mL
       152,00 €
    Marketsource
    Marketsource