Order Entry
Belgium
Orders LinkContactUsLinkComponent
ALDEFLUOR™ Assay Buffer

ALDEFLUOR™ Assay Buffer

Additional assay buffer for use with the ALDEFLUOR™ kit, 55 mL
Catalog # STMC01702
Marketsource
ALDEFLUOR™ Assay Buffer
Catalog # STMC01702
Marketsource
CAS Number:  
ALDEFLUOR™ Assay Buffer
ALDEFLUOR™ Assay Buffer

Additional assay buffer for use with the ALDEFLUOR™ kit, 55 mL

CAS Number:  
Price
229,00 €
Log in for availability
1 * 55 mL
Product Details & Documents

Use ALDEFLUOR™ Assay Buffer to optimize the detection of aldehyde dehydrogenase-bright (ALDHbr) cells and maximize fluorescent signal stability during the ALDEFLUOR™ assay.

ALDEFLUOR™ Assay Buffer is recommended for use in all procedures performed after ALDH staining to inhibit the active efflux of the ALDEFLUOR™ reaction product and to avoid loss in the assay signal.

Application: Cancer, epithelial cell biology, neuroscience, stem cell biology
Species: Human, mouse, non human primate, rat and others.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    55 mL
  • Description:
    Additional assay buffer for use with the ALDEFLUOR™ kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.