RoboSep™ Buffer
About this item
RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.
Application: Immunology
Species: Human, mouse, non human primate, other, rat
Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
2 Options Available Below
Product Details & Documents
RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.
Application: Immunology
Species: Human, mouse, non human primate, other, rat
Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



