Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant Shh (C24II)
Human Recombinant Shh (C24II)
Certificates
undefined
Human Recombinant Shh (C24II)

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Shh (C24II)
  • Purity:
    ≥95%

Specifications

Frequently Bought Together