Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant Noggin
Mouse Recombinant Noggin
Certificates
undefined
Mouse Recombinant Noggin

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein synonyms:
    SYM1|SYNS1|NOG
  • Protein/peptide name:
    Noggin
  • Purity:
    ≥95%

Specifications

Frequently Bought Together