Order Entry
Belgium
ContactUsLinkComponent
Mouse Recombinant LIF
Mouse Recombinant LIF
Certificates
undefined
Mouse Recombinant LIF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    LIF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together