Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mouse Recombinant PDGF-BB

Mouse Recombinant PDGF-BB

Marketsource
Mouse Recombinant PDGF-BB
Marketsource
Certificates
Mouse Recombinant PDGF-BB

2 Options Available Below

Product Details & Documents

The platelet-derived growth factor (PDGF) family has five heparin-binding members that assemble into four homodimers (PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD) and one heterodimer (PDGF-AB; Fretto et al.; Li and Eriksson). PDGF signals through the receptor tyrosine kinases PDGFRα and PDGFRβ. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). PDGF-BB is secreted by osteoblasts to induce mesenchymal stem cell migration and angiogenesis. It has also been shown that PDGF-BB is secreted by preosteoclasts during bone modeling and remodeling to induce angiogenesis and thus proper osteogenesis (Xie et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78178
  • STMC78178.1
  • Protein/peptide name:
  • PDGF-BB
  • PDGF-BB
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Mouse
  • Mouse
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 190,00 €
    190,00 €
    1 * 1 EA
       190,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78178
    Protein/peptide name
    PDGF-BB
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Purity
    ≥95%
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 755,00 €
    755,00 €
    1 * 1 EA
       755,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78178.1
    Protein/peptide name
    PDGF-BB
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Purity
    ≥95%
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.