Order Entry
Belgium
ContactUsLinkComponent
 
Human Recombinant FGF-16
 
undefined
Human Recombinant FGF-16

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    FGF-16
  • Purity:
    ≥95%

 

Frequently Bought Together