Order Entry
Belgium
ContactUsLinkComponent
Human/Mouse Recombinant TGF-beta 3, ACF
Human/Mouse Recombinant TGF-beta 3, ACF
Certificates
undefined
Human/Mouse Recombinant TGF-beta 3, ACF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Protein/peptide name:
    TGF-beta 3, ACF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together