Order Entry
Belgium
ContactUsLinkComponent
Mouse/Rat Recombinant RANTES (CCL5)
Mouse/Rat Recombinant RANTES (CCL5)
Certificates
undefined
Mouse/Rat Recombinant RANTES (CCL5)

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
    Rat
  • Protein/peptide name:
    RANTES (CCL5)
  • Purity:
    ≥95%

Specifications

Frequently Bought Together