Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant IFN-alpha 2A
Human Recombinant IFN-alpha 2A
Certificates
undefined
Human Recombinant IFN-alpha 2A

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IFN-alpha 2A
  • Purity:
    ≥95%

Specifications

Frequently Bought Together