Order Entry
Belgium
ContactUsLinkComponent
 
Mouse Recombinant IL-5
 
undefined
Mouse Recombinant IL-5

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-5
  • Purity:
    ≥95%

 

Frequently Bought Together