Order Entry
Belgium
ContactUsLinkComponent
Rat Recombinant TNF-alpha
Rat Recombinant TNF-alpha
Catalog # STMC78124.1
CAS Number:  
undefined
Rat Recombinant TNF-alpha
Catalog # STMC78124.1
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Seq:
    1202
  • Protein/peptide name:
    TNF-alpha
  • Purity:
    ≥95%

Specifications

About this item

Tumor necrosis factor-alpha (TNF-α) is a pro-inflammatory cytokine that activates NF-κB, MAPK, and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts. TNF-α has cytotoxic effects on cancerous cells by stimulating anti-tumor immunosuppressive responses. TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová and Hošek). Other effects of TNF-α include vasodilatation and edema formation. In vitro studies of adult rat neural progenitor cells (NPCs) demonstrate that TNF-α reduces neurogenesis in dentate gyrus-derived NPCs, and promotes astrogliogenesis in subventricular zone-derived NPCs (Borsini et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').