Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant Apolipoprotein A-I, His tag
Human Recombinant Apolipoprotein A-I, His tag
Catalog # STMC100-1322
CAS Number:  
undefined
Human Recombinant Apolipoprotein A-I, His tag
Catalog # STMC100-1322
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Apolipoprotein A1
  • Protein/peptide name:
    Apolipoprotein A-I
  • Purity:
    ≥92%

Specifications

About this item

A member of the apolipoprotein family, Apolipoprotein A-I (apo A-I) is a major glycoprotein component of high density lipoprotein (HDL) particles (Frank and Marcel) which facilitate the reverse transport of cholesterol from peripheral tissues to the liver. Apo A-I binds to the scavenger receptor SR-A1, allowing HDL particles to interact with cells in peripheral tissues (Neyen et al.), and is involved in the esterification of cholesterol, acting as a cofactor for lecithin cholesterol acyltransferase (LCAT; Frohlich). Apo A-I has been shown to bind to the scavenger receptor SR-B1 (Williams et al.) and the beta chain of ATP synthase (Martinez et al.) on hepatocytes, highlighting its importance in HDL endocytosis. Mutations in the APOA1 encoding gene may result in dysregulated HDL levels, leading to an increased risk of amyloidosis and ischemic heart disease (Haase et al.; Obici et al.). This protein product contains a His-residue tag at the amino end of the polypeptide chain. This protein product contains a His-residue tag at the amino end of the polypeptide chain. For consistency and reproducibility across your applications, sclerostin from STEMCELL comes lyophilised with ≥92% purity.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').