Orders LinkContactUsLinkComponent
Water Separator
Water Separator
Water Separator
Certificates
4 Options Available Below
Product Details & Documents
Water separator made of Borosilicate glass 3.3.
  • Graduation 10 ml in 1/10, 0-point on the stopcock
  • With 2-way tap
Product Family Options
Product Information
  • Capacity
    Graduation
    Cone to condenser
    Cone to flask
    Availability
    Price
  • NEUB-90-10292914
    Water separator, tap NS 14,5/1,5 mm
    Water separator, tap NS 14,5/1,5 mm
    10 ml
    0,1 ml
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 513,00 €
    513,00 €
    1 * 1 ST
       513,00 €
  • NEUB-90-10292919
    Water separator, tap NS 18,8/2,5 mm
    Water separator, tap NS 18,8/2,5 mm
    10 ml
    0,1 ml
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 528,00 €
    528,00 €
    1 * 1 ST
       528,00 €
  • NEUB03829010292914
    Water separator, two way stopcock, NS 14,5/1,5 mm
    Water separator, two way stopcock, NS 14,5/1,5 mm
    10 ml
    0,1 ml
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 503,00 €
    503,00 €
    1 * 1 ST
       503,00 €
  • NEUB03829010292919
    Water separator, two way stopcock, NS 18,8/2,5 mm
    Water separator, two way stopcock, NS 18,8/2,5 mm
    10 ml
    0,1 ml
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 518,00 €
    518,00 €
    1 * 1 ST
       518,00 €